Assay Details
Gene Symbol: Fam150b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109372.1 | NP_001102842.1 | 4 - 5 | 893 | 102 | |
XM_006239971.2 | XP_006240033.1 | 3 - 4 | 1033 | 102 | ||
GenBank mRNA | BC167018.1 | - | 4 - 5 | 934 | 7307720 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
