Assay Details
Gene Symbol: Galm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007704.1 | NP_001007705.1 | 5 - 6 | 828 | 113 | |
GenBank mRNA | BC081876.1 | - | 5 - 6 | 828 | 7107751 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
