Assay Details
Gene Symbol: Rdh12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108037.1 | NP_001101507.1 | 2 - 3 | 325 | 59 | |
XM_008764779.1 | XP_008763001.1 | 1 - 2 | 455 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
