Assay Details
Gene Symbol: Arg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019168.1 | NP_062041.1 | 6 - 7 | 820 | 100 | |
GenBank mRNA | U90887.1 | - | 6 - 7 | 820 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
