Assay Details
Gene Symbol: Ankmy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108019.1 | NP_001101489.1 | 5 - 6 | 720 | 139 | |
XM_006240042.2 | XP_006240104.1 | 5 - 6 | 3105 | 139 | ||
XM_006240043.2 | XP_006240105.2 | 6 - 7 | 3422 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
