Assay Details
Gene Symbol: Ankmy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108019.1 | NP_001101489.1 | 8 - 9 | 1200 | 80 | |
XM_006240042.2 | XP_006240104.1 | 9 - 10 | 3714 | 80 | ||
XM_006240043.2 | XP_006240105.2 | 10 - 11 | 4031 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P573958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396892 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97981 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P573958 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P573958 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P573958 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P138185 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97954 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10884268 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884265 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884266 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884267 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884273 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884270 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884272 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884269 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10884264 | - | - |
Rat Genome 230 2.0 Array | 1379702_at | - | - |
Rat Genome 230 2.0 Array | 1372742_at | - | - |
Rat Genome 230 2.0 Array | 1379702_PM_at | - | - |
Rat Genome 230 2.0 Array | 1372742_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379702_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1372742_at | - | - |
Affymetrix HT Rat Focus Array | 1372742_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379702_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600004451.rn.1 | TC0600000626.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600004450.rn.1 | TC0600000626.rn.1 | RTA TC |
Rat Expression Set 230 | 1372742_at | - | - |
Rat Expression Set 230 | 1372742_PM_at | - | - |
Rat Expression Set 230 | 1379702_at | - | - |
Rat Expression Set 230 | 1379702_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10884264 | - | - |
Rat Genome U34 Set | rc_AA892168_at | - | - |
Rat Genome U34 Set | rc_AI029529_at | - | - |
Rat Genome U34 Set | rc_AI179316_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21322163 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363684 | - | - |
