Assay Details
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185025.1 | NP_001171954.1 | 3 - 4 | 304 | 131 | |
XM_008764655.1 | XP_008762877.1 | 3 - 4 | 259 | 131 | ||
GenBank mRNA | BP486352.1 | - | 3 - 4 | 310 | 132 | |
CO399813.1 | - | 3 - 4 | 280 | 131 | ||
FQ210674.1 | - | 3 - 4 | 322 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
