Assay Details
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006239906.2 | 7 - 8 | 1027 | 84 | ||
XM_006239907.2 | XP_006239969.1 | 7 - 8 | 1017 | 84 | ||
XM_006239908.2 | XP_006239970.1 | 6 - 7 | 933 | 84 | ||
XM_008764573.1 | 7 - 8 | 1000 | 84 | |||
GenBank mRNA | BC078882.1 | - | 5 - 6 | 705 | 7115603 | 84 |
J04791.1 | - | 7 - 8 | 821 | 84 | ||
M16982.1 | - | 7 - 8 | 1085 | 84 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
