Assay Details
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_233975.8 | XP_233975.5 | 3 - 4 | 463 | 145 | |
GenBank mRNA | FQ224595.1 | - | 3 - 4 | 432 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
