Assay Details
Gene Symbol: Bre | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199270.1 | NP_954891.1 | 6 - 7 | 646 | 65 | |
XM_006239802.2 | XP_006239864.1 | 7 - 8 | 755 | 65 | ||
GenBank mRNA | BC061573.1 | - | 6 - 7 | 646 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
