Assay Details
Gene Symbol: Zp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172330.3 | NP_758833.2 | 1 - 2 | 218 | 80 | |
GenBank mRNA | AF456325.1 | - | 1 - 2 | 219 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
