Assay Details
Gene Symbol: Gosr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053584.2 | NP_446036.1 | 3 - 4 | 237 | 99 | |
XM_006246914.1 | XP_006246976.1 | 3 - 4 | 305 | 99 | ||
XM_008768132.1 | XP_008766354.1 | 2 - 3 | 108 | 99 | ||
GenBank mRNA | BC126068.1 | - | 3 - 4 | 241 | 7387185 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768132.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
