Assay Details
Gene Symbol: Gosr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053584.2 | NP_446036.1 | 7 - 8 | 551 | 82 | |
XM_006246914.1 | XP_006246976.1 | 7 - 8 | 619 | 82 | ||
XM_008768132.1 | XP_008766354.1 | 6 - 7 | 422 | 82 | ||
GenBank mRNA | BC126068.1 | - | 7 - 8 | 555 | 7387185 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
