Assay Details
Gene Symbol: Slc25a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007674.1 | NP_001007675.1 | 3 - 4 | 359 | 103 | |
GenBank mRNA | BC079002.1 | - | 3 - 4 | 359 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
