Assay Details
Gene Symbol: Anapc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247906.2 | XP_006247968.1 | 1 - 2 | 219 | 85 | |
GenBank mRNA | FQ228975.1 | - | 1 - 2 | 74 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006247906.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
