Assay Details
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031149.1 | NP_112411.1 | 11 - 12 | 1184 | 87 | |
GenBank mRNA | AB000491.1 | - | 11 - 12 | 1184 | 87 | |
BC058462.1 | - | 11 - 12 | 1180 | 6891005 | 87 | |
D83521.1 | - | 11 - 12 | 1202 | 87 | ||
FQ213263.1 | - | 11 - 12 | 1201 | 87 | ||
FQ233831.1 | - | 11 - 12 | 1187 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
