Assay Details
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031149.1 | NP_112411.1 | 1 - 2 | 40 | 136 | |
GenBank mRNA | AB000491.1 | - | 1 - 2 | 40 | 136 | |
BC058462.1 | - | 1 - 2 | 36 | 6891005 | 136 | |
D83521.1 | - | 1 - 2 | 58 | 136 | ||
FQ213735.1 | - | 1 - 2 | 32 | 136 | ||
FQ233831.1 | - | 1 - 2 | 43 | 136 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB000491.1, BC058462.1, D83521.1, NM_031149.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
