Assay Details
Gene Symbol: Il12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022611.1 | NP_072133.1 | 4 - 5 | 690 | 71 | |
XM_006246147.2 | XP_006246209.1 | 5 - 6 | 899 | 71 | ||
GenBank mRNA | AF133197.1 | - | 4 - 5 | 690 | 71 | |
U16674.1 | - | 4 - 5 | 706 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
