Assay Details
Gene Symbol: Galk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008282.1 | NP_001008283.1 | 2 - 3 | 384 | 71 | |
GenBank mRNA | BC085919.1 | - | 2 - 3 | 384 | 71 | |
FQ219172.1 | - | 2 - 3 | 398 | 71 | ||
FQ234739.1 | - | 2 - 3 | 357 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
