Assay Details
Gene Symbol: Galk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008282.1 | NP_001008283.1 | 5 - 6 | 820 | 58 | |
GenBank mRNA | BC085919.1 | - | 5 - 6 | 820 | 58 | |
FQ234739.1 | - | 5 - 6 | 793 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
