Assay Details
Gene Symbol: Ube2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031138.2 | NP_112400.1 | 2 - 3 | 230 | 160 | |
XM_006246343.2 | XP_006246405.1 | 3 - 4 | 255 | 160 | ||
GenBank mRNA | BC070946.1 | - | 2 - 3 | 223 | 7103580 | 160 |
DN931313.1 | - | 2 - 3 | 260 | 160 | ||
FQ229894.1 | - | 2 - 3 | 241 | 160 | ||
M62388.1 | - | 2 - 3 | 172 | 160 |
Gene Symbol: LOC103694902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008773893.1 | XP_008772115.1 | 3 - 4 | 254 | 160 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
