Assay Details
Gene Symbol: Myh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135157.1 | NP_001128629.1 | 5666 | 158 | ||
GenBank mRNA | FQ215961.1 | - | 5 - 6 | 764 | 158 | |
L13606.1 | - | 3 - 4 | 301 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P105383 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P109919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25899 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109919 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P105383 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109919 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P105383 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P105383 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P109919 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P179996 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1031380 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10734544 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734546 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734549 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734543 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734539 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734555 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734532 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734537 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734551 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734542 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734538 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734545 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734553 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734547 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734531 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734554 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734550 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734536 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734540 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734548 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734533 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734552 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734541 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734534 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734535 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10734530 | - | - |
Rat Genome 230 2.0 Array | 1388139_at | - | - |
Rat Genome 230 2.0 Array | 1388139_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388139_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388139_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1000007207.rn.1 | TC1000000862.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000007209.rn.1 | TC1000000862.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000007208.rn.1 | TC1000000862.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1000007206.rn.1 | TC1000000862.rn.1 | RTA TC |
Rat Expression Set 230 | 1388139_at | - | - |
Rat Expression Set 230 | 1388139_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10734530 | - | - |
Rat Genome U34 Set | L13606_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21319732 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371839 | - | - |
