Assay Details
Gene Symbol: Birc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022274.1 | NP_071610.1 | 2 - 3 | 221 | 147 | |
GenBank mRNA | AF276775.1 | - | 2 - 3 | 221 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
