Assay Details
Gene Symbol: Lasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032613.2 | NP_116002.1 | 3 - 4 | 333 | 70 | |
XM_006247288.2 | XP_006247350.1 | 3 - 4 | 265 | 70 | ||
GenBank mRNA | AF242187.1 | - | 3 - 4 | 247 | 70 | |
BC099791.1 | - | 3 - 4 | 333 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
