Assay Details
Gene Symbol: Shbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012650.1 | NP_036782.1 | 6 - 7 | 887 | 86 | |
XM_006246586.2 | XP_006246648.1 | 7 - 8 | 1636 | 86 | ||
XM_006246592.2 | XP_006246654.1 | 6 - 7 | 994 | 86 | ||
XM_008767771.1 | XP_008765993.1 | 7 - 8 | 1101 | 86 | ||
XM_008767772.1 | XP_008765994.1 | 6 - 7 | 909 | 86 | ||
GenBank mRNA | BC097336.1 | - | 6 - 7 | 875 | 86 | |
M15034.1 | - | 6 - 7 | 887 | 86 | ||
M31179.1 | - | 6 - 7 | 839 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
