Assay Details
Gene Symbol: Pdpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031081.1 | NP_112343.1 | 3 - 4 | 394 | 148 | |
XM_006246036.2 | XP_006246098.1 | 3 - 4 | 2246 | 148 | ||
XM_006246038.2 | XP_006246100.1 | 3 - 4 | 456 | 148 | ||
GenBank mRNA | Y15748.1 | - | 3 - 4 | 394 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
