Assay Details
Gene Symbol: Slc25a39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024792.1 | NP_001019963.1 | 2 - 3 | 196 | 140 | |
GenBank mRNA | BC097349.1 | - | 2 - 3 | 196 | 140 | |
FQ228904.1 | - | 2 - 3 | 186 | 140 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC097349.1, NM_001024792.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
