Assay Details
Gene Symbol: Slc25a39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024792.1 | NP_001019963.1 | 7 - 8 | 626 | 84 | |
XM_006247374.2 | XP_006247436.1 | 7 - 8 | 964 | 84 | ||
XM_006247376.2 | XP_006247438.1 | 7 - 8 | 649 | 84 | ||
GenBank mRNA | BC097349.1 | - | 7 - 8 | 626 | 84 | |
FQ228904.1 | - | 7 - 8 | 616 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
