Assay Details
Gene Symbol: Slc25a39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024792.1 | NP_001019963.1 | 9 - 10 | 912 | 119 | |
XM_006247374.2 | XP_006247436.1 | 9 - 10 | 1250 | 119 | ||
XM_006247376.2 | XP_006247438.1 | 9 - 10 | 935 | 119 | ||
GenBank mRNA | BC081862.1 | - | 9 - 10 | 829 | 119 | |
BC097349.1 | - | 9 - 10 | 912 | 119 | ||
FQ228904.1 | - | 9 - 10 | 904 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
