Assay Details
Gene Symbol: Pmp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017037.1 | NP_058733.1 | 3 - 4 | 383 | 57 | |
XM_006246584.2 | XP_006246646.1 | 3 - 4 | 381 | 57 | ||
XM_006246585.2 | XP_006246647.1 | 3 - 4 | 390 | 57 | ||
GenBank mRNA | M69139.1 | - | 3 - 4 | 299 | 57 | |
S55427.1 | - | 3 - 4 | 299 | 57 | ||
X62431.1 | - | 3 - 4 | 383 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
