Assay Details
Gene Symbol: P2rx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080780.2 | NP_542958.2 | 11 - 12 | 1312 | 104 | |
XM_006246566.2 | XP_006246628.1 | 10 - 11 | 1320 | 104 | ||
GenBank mRNA | X92069.1 | - | 11 - 12 | 1312 | 104 | |
X97328.1 | - | 11 - 12 | 1178 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
