Assay Details
Gene Symbol: P2rx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080780.2 | NP_542958.2 | 2 - 3 | 466 | 67 | |
GenBank mRNA | X92069.1 | - | 2 - 3 | 466 | 67 | |
X97328.1 | - | 2 - 3 | 332 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
