Assay Details
Gene Symbol: Dcxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134387.1 | NP_599214.1 | 5 - 6 | 465 | 70 | |
XM_008768465.1 | XP_008766687.1 | 5 - 6 | 511 | 70 | ||
XM_008768466.1 | XP_008766688.1 | 5 - 6 | 511 | 70 | ||
XM_008768467.1 | XP_008766689.1 | 5 - 6 | 511 | 70 | ||
GenBank mRNA | AB061719.1 | - | 5 - 6 | 465 | 70 | |
FQ233401.1 | - | 5 - 6 | 464 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
