Assay Details
Gene Symbol: Pctp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247124.2 | XP_006247186.1 | 5 - 6 | 568 | 108 | |
XM_008767982.1 | XP_008766204.1 | 7 - 8 | 789 | 108 | ||
XM_008767983.1 | XP_008766205.1 | 7 - 8 | 743 | 108 | ||
GenBank mRNA | AF040261.1 | - | 5 - 6 | 526 | 108 | |
BC078854.1 | - | 5 - 6 | 587 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
