Assay Details
Gene Symbol: Zpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007011.1 | NP_001007012.1 | 2 - 3 | 215 | 169 | |
XM_006247384.2 | XP_006247446.1 | 2 - 3 | 1345 | 169 | ||
XM_006247385.2 | XP_006247447.1 | 2 - 3 | 1272 | 169 | ||
XM_006247387.2 | XP_006247449.1 | 2 - 3 | 1438 | 169 | ||
GenBank mRNA | AY251605.1 | - | 2 - 3 | 207 | 169 | |
BC082007.1 | - | 2 - 3 | 215 | 7131668 | 169 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006247384.2, XM_006247385.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
