Assay Details
Gene Symbol: Zpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007011.1 | NP_001007012.1 | 3 - 4 | 346 | 80 | |
XM_006247384.2 | XP_006247446.1 | 3 - 4 | 1476 | 80 | ||
XM_006247385.2 | XP_006247447.1 | 3 - 4 | 1403 | 80 | ||
XM_006247387.2 | XP_006247449.1 | 3 - 4 | 1569 | 80 | ||
GenBank mRNA | AY251605.1 | - | 3 - 4 | 338 | 80 | |
AY251606.1 | - | 2 - 3 | 272 | 80 | ||
BC082007.1 | - | 3 - 4 | 346 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
