Assay Details
Gene Symbol: Zpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007011.1 | NP_001007012.1 | 4 - 5 | 509 | 98 | |
XM_006247384.2 | XP_006247446.1 | 4 - 5 | 1639 | 98 | ||
XM_006247385.2 | XP_006247447.1 | 4 - 5 | 1566 | 98 | ||
GenBank mRNA | AY251605.1 | - | 4 - 5 | 501 | 98 | |
AY251606.1 | - | 3 - 4 | 435 | 98 | ||
BC082007.1 | - | 4 - 5 | 509 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
