Assay Details
Gene Symbol: Ttll6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768194.1 | XP_008766416.1 | 1 - 2 | 110 | 69 | |
XM_008768195.1 | XP_008766417.1 | 1 - 2 | 110 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768194.1, XM_008768195.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
