Assay Details
Gene Symbol: Ttll6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768194.1 | XP_008766416.1 | 4 - 5 | 541 | 97 | |
XM_008768195.1 | XP_008766417.1 | 4 - 5 | 541 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
