Assay Details
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001718.1 | NP_001001718.1 | 7 - 8 | 1052 | 62 | |
XM_006246639.2 | XP_006246701.1 | 8 - 9 | 1252 | 62 | ||
XM_006246640.1 | XP_006246702.1 | 6 - 7 | 561 | 62 | ||
GenBank mRNA | AY623900.1 | - | 7 - 8 | 1052 | 62 | |
BC158569.1 | - | 8 - 9 | 1255 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
