Assay Details
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001718.1 | NP_001001718.1 | 1 - 2 | 312 | 109 | |
XM_006246639.2 | XP_006246701.1 | 2 - 3 | 506 | 109 | ||
XM_006246640.1 | XP_006246702.1 | 1 - 2 | 42 | 109 | ||
GenBank mRNA | AY623900.1 | - | 1 - 2 | 312 | 109 | |
BC158569.1 | - | 2 - 3 | 509 | 9101477 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006246640.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
