Assay Details
Gene Symbol: Atpaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107006.1 | NP_001100476.1 | 4 - 5 | 583 | 121 | |
GenBank mRNA | FQ213754.1 | - | 4 - 5 | 573 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
