Assay Details
Gene Symbol: Hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033349.2 | NP_203500.2 | 1 - 2 | 112 | 141 | |
GenBank mRNA | BC097301.1 | - | 1 - 2 | 64 | 141 | |
CB715597.1 | - | 1 - 2 | 141 | 141 | ||
FQ229806.1 | - | 1 - 2 | 181 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
