Assay Details
Gene Symbol: Hagh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033349.2 | NP_203500.2 | 8 - 9 | 867 | 78 | |
XM_006245899.1 | XP_006245961.1 | 8 - 9 | 844 | 78 | ||
XM_006245900.1 | XP_006245962.1 | 7 - 8 | 758 | 78 | ||
GenBank mRNA | BC097301.1 | - | 8 - 9 | 819 | 78 | |
FQ229806.1 | - | 8 - 9 | 936 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
