Assay Details
Gene Symbol: Sat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108278.1 | NP_001101748.1 | 2 - 3 | 265 | 90 | |
XM_006246743.1 | XP_006246805.1 | 3 - 4 | 386 | 90 | ||
XM_006246744.2 | XP_006246806.1 | 3 - 4 | 287 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
