Assay Details
Gene Symbol: Faxdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001077260.4 | XP_001077260.4 | 6 - 7 | 826 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
