Assay Details
Gene Symbol: Aldoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012497.1 | NP_036629.1 | 3 - 4 | 436 | 125 | |
GenBank mRNA | BC059154.1 | - | 3 - 4 | 411 | 125 | |
BC099749.1 | - | 3 - 4 | 422 | 125 | ||
FQ213506.1 | - | 3 - 4 | 439 | 125 | ||
FQ214420.1 | - | 3 - 4 | 462 | 125 | ||
FQ228893.1 | - | 3 - 4 | 436 | 125 | ||
X06984.1 | - | 3 - 4 | 436 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
