Assay Details
Gene Symbol: Aldoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012497.1 | NP_036629.1 | 4 - 5 | 486 | 59 | |
GenBank mRNA | BC099749.1 | - | 4 - 5 | 472 | 59 | |
FQ213506.1 | - | 4 - 5 | 489 | 59 | ||
FQ214420.1 | - | 4 - 5 | 512 | 59 | ||
FQ228893.1 | - | 4 - 5 | 486 | 59 | ||
X06984.1 | - | 4 - 5 | 486 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
