Assay Details
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012751.1 | NP_036883.1 | 5 - 6 | 700 | 126 | |
XM_006246596.2 | XP_006246658.1 | 6 - 7 | 673 | 126 | ||
GenBank mRNA | BC085757.1 | - | 5 - 6 | 745 | 126 | |
D28561.1 | - | 5 - 6 | 700 | 126 | ||
FQ216450.1 | - | 6 - 7 | 639 | 126 | ||
FQ232657.1 | - | 5 - 6 | 126 | |||
FQ233419.1 | - | 5 - 6 | 735 | 126 | ||
J04524.1 | - | 5 - 6 | 682 | 126 | ||
KJ696745.1 | - | 5 - 6 | 585 | 126 | ||
M25482.1 | - | 5 - 6 | 665 | 126 | ||
X14771.1 | - | 5 - 6 | 713 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
