Assay Details
Gene Symbol: Srpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108243.2 | NP_001101713.1 | 2 - 3 | 139 | 135 | |
GenBank mRNA | BC168975.1 | - | 2 - 3 | 139 | 135 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168975.1, NM_001108243.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
